2,3,5,6-Tetramethylbenzenesulphonyl chloride ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,3,5,6-Tetramethylbenzene-1-sulphonyl chloride
A13313.09 A13313.18
A13313.09EA 1025 SEK
A13313.09 A13313.18
2,3,5,6-Tetramethylbenzenesulphonyl chloride ≥98%
2,3,5,6-Tetramethylbenzenesulphonyl chloride
Purity: 98% CAS No.: 60706-63-0 Molecular Formula: C10H13ClO2S Formula Weight: 232.73 Melting Point: 97-101° Beilstein Reference: 1876644 UN No.: 3261 MDL No.: MFCD00014723
Formel: C₁₀H₁₃ClO₂S
MW: 232,73 g/mol
Smältpunkt: 97…101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014723
CAS nummer: 60706-63-0
UN: 3261
ADR: 8,II