2,3,5-Trimetylhydrokinon 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,3,5-Trimethylbenzene-1,4-diol, Trimethylhydroquinone
140151000 140155000
ACRO140151000EA 1125 SEK
ACRO140151000 ACRO140155000
2,3,5-Trimetylhydrokinon 97%
2,3,5-Trimetylhydrokinon
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokpunkt: 295 °C (1013 hPa)
Smältpunkt: 169…174 °C
Flampunkt: 191 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002346
CAS nummer: 700-13-0
EINECS: 211-838-3
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White to beige to orange Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 174 °C
GC ≥96.0 %