2,4,6-Trimethylbenzoyl chloride ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A14407.09 A14407.18
A14407.09EA 1800 SEK
A14407.09 A14407.18
2,4,6-Trimethylbenzoyl chloride ≥98%
2,4,6-Trimethylbenzoyl chloride
Purity: 98+% CAS No.: 938-18-1 Molecular Formula: C10H11ClO Formula Weight: 182.65 Flash Point: 110°(230°F) Refractive Index: 1.5290 Boiling Point: 143-146°/60mm Beilstein Reference: 776108 UN No.: 3265 MDL No.: MFCD00013650
Formel: C₁₀H₁₁ClO
Kokpunkt: 143…146 °C (60 mmHg)
Densitet: 1,10
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013650
CAS nummer: 938-18-1
EINECS: 213-339-6
UN: 3265
ADR: 8,II