4-Methylbenzophenone ≥98%
Leverantör: Thermo Scientific
Synonyms: p-Methylbenzophenone, Phenyl(p-tolyl)methanone
A15479.18 A15479.36
A15479.18EA 1300 SEK
A15479.18 A15479.36
4-Methylbenzophenone ≥98%
4-Methylbenzophenone
Purity: 98% CAS No.: 134-84-9 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 143°(289°F) Melting Point: 53-57° Boiling Point: 326° Beilstein Reference: 1909310 Merck Reference: 14,7317 MDL No.: MFCD00008553
Formel: C₁₄H₁₂O
MW: 196,25 g/mol
Kokpunkt: 326 °C
Smältpunkt: 53…57 °C
Densitet: 0,9926 g/cm³ (20 °C)
Flampunkt: 143 °C (289 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008553
CAS nummer: 134-84-9
EINECS: 205-159-1
Merck Index: 14,7317