2-(Hydroxymethyl)benzyl alcohol 97%
Leverantör: Thermo Scientific
Synonyms: o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol
180410025 180410100
ACRO180410025EA 1575 SEK
ACRO180410025 ACRO180410100
2-(Hydroxymethyl)benzyl alcohol 97%
2-(Hydroxymethyl)benzyl alcohol
Formel: C₈H₁₀O₂
MW: 138,17 g/mol
Kokpunkt: 291 °C (760 mmHg)
Smältpunkt: 61…64 °C
Flampunkt: 145 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004626
CAS nummer: 612-14-6
EINECS: 210-293-9
Order Now
Specifikation testresultat
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 61.0 to 65.0 °C
GC ≥96.0 %