4-Methylbenzyl alcohol ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-(Hydroxymethyl)toluole, p-Tolylmethanol, (4-Methylphenyl)methanol
A15315.14 A15315.22 A15315.36
A15315.14EA 1050 SEK
A15315.14 A15315.22 A15315.36
4-Methylbenzyl alcohol ≥99%
4-Methylbenzyl alcohol
Purity: 99% CAS No.: 589-18-4 Molecular Formula: C8H10O Formula Weight: 122.17 Melting Point: 57-61° Boiling Point: 217° MDL No.: MFCD00004664
Formel: C₈H₁₀O
MW: 122,17 g/mol
Kokpunkt: 217 °C
Smältpunkt: 57…61 °C
Densitet: 0,978
Storage Temperature: Omgivande
MDL Number: MFCD00004664
CAS nummer: 589-18-4
EINECS: 209-639-1