3-Methylbenzylamine 98%
Leverantör: Thermo Scientific
Fara
Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene
155480010 155480050 155480250
ACRO155480010EA 445 SEK
ACRO155480010 ACRO155480050 ACRO155480250
3-Methylbenzylamine 98%
3-Methylbenzylamine
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokpunkt: 202…205 °C (760 mmHg)
Densitet: 0,966 g/cm³
Flampunkt: 80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008118
CAS nummer: 100-81-2
EINECS: 202-890-8
UN: 2735
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5380 (20°C, 589 nm)
GC ≥97.5 %