2,5-Xylyl isothiocyanate

Leverantör: Thermo Scientific

Synonyms: 2,5-Dimethylphenylisothiocyanate, 2,5-Dimethylphenyl isothiocyanate

SB01618EA
MAYBSB01618EAEA 0 SEK
MAYBSB01618EA
2,5-Xylyl isothiocyanate
2,5-Xylyl isothiocyanate
Formel: C₉H₉NS
MW: 163,24 g/mol
MDL Number: MFCD00046798
CAS nummer: 19241-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR