2,4-Xylyl isothiocyanate

Leverantör: Thermo Scientific

Synonyms: 2,4-Dimethylphenylisothiocyanate, 2,4-Dimethylphenyl isothiocyanate

SB01620EA
MAYBSB01620EAEA 0 SEK
MAYBSB01620EA
2,4-Xylyl isothiocyanate
2,4-Xylyl isothiocyanate
Formel: C₉H₉NS
MW: 163,24 g/mol
Kokpunkt: 262…263 °C (1013 hPa)
Smältpunkt: 31,5 °C
MDL Number: MFCD00022054
CAS nummer: 39842-01-8
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR