α,α,α,α',α',α'-Hexafluoro-2,5-xylidine ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,5-Bis(trifluoromethyl)aniline
L14706.03 L14706.06 L14706.14
L14706.03EA 895 SEK
L14706.03 L14706.06 L14706.14
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine ≥99%
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
Purity: 99% CAS No.: 328-93-8 Molecular Formula: C8H5F6N Formula Weight: 229.13 Flash Point: 71°(159°F) Refractive Index: 1.4338 Odour: Recognizable Boiling Point: 70-71°/15mm Beilstein Reference: 2653046 UN No.: 2810 MDL No.: MFCD00074940
Formel: C₈H₅F₆N
MW: 229,12 g/mol
Kokpunkt: 70…71 °C (15 mmHg)
Densitet: 1,467
Flampunkt: 71 °C (159 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00074940
CAS nummer: 328-93-8
UN: 2810
ADR: 6.1,III