3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1,3-Bis(trifluoromethyl)-5-isothiocyanatobenzene, 1-Isothiocyanato-3,5-bis(trifluoromethyl)benzene
B20206.03 B20206.06 B20206.14
B20206.03EA 770 SEK
B20206.03 B20206.06 B20206.14
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate
Purity: 98% CAS No.: 23165-29-9 Molecular Formula: C9H3F6NS Formula Weight: 271.19 Flash Point: 108°(226°F) Refractive Index: 1.500 Odour: Irritating Boiling Point: 109-110°/35mm Beilstein Reference: 2990816 UN No.: 2922 MDL No.: MFCD00040838
Formel: C₉H₃F₆NS
MW: 271.19 g/mol
Kokpunkt: 109…110 °C (35 mmHg)
Densitet: 1.485
Flampunkt: 108 °C (226 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00040838
CAS nummer: 23165-29-9
ADR: 8 III