4'-(Trifluoromethyl)acetophenone ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone
A15529.06 A15529.14
A15529.06EA 1325 SEK
A15529.06 A15529.14
4'-(Trifluoromethyl)acetophenone ≥98%
4'-(Trifluoromethyl)acetophenone
Purity: 98+% CAS No.: 709-63-7 Molecular Formula: C9H7F3O Formula Weight: 188.15 Flash Point: 84°(183°F) Refractive Index: 1.4620 Odour: Characteristic Melting Point: 29-35° Boiling Point: 83-85°/9mm Beilstein Reference: 1870425 MDL No.: MFCD00000401
Formel: C₉H₇F₃O
Kokpunkt: 83…85 °C (9 mmHg)
Smältpunkt: 29…35 °C
Densitet: 0,924
Flampunkt: 84 °C (183 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000401
CAS nummer: 709-63-7
EINECS: 211-913-0