4-Bromo-2,6-xylidine 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Bromo-2,6-dimethylaniline
185190050 185190250
ACRO185190050EA 440 SEK
ACRO185190050 ACRO185190250
4-Bromo-2,6-xylidine 98%
4-Bromo-2,6-xylidine
Formel: C₈H₁₀BrN
MW: 200.08 g/mol
Kokpunkt: 120 °C (0.2 mmHg)
Smältpunkt: 48…52 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007826
CAS nummer: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance Beige to brown to purple Powder or crystals
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %