1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene
A12337.09 A12337.18 A12337.30
A12337.09EA 1100 SEK
A12337.09 A12337.18 A12337.30
1-Bromo-3,5-bis(trifluoromethyl)benzene ≥98%
1-Bromo-3,5-bis(trifluoromethyl)benzene
Purity: 98% CAS No.: 328-70-1 Molecular Formula: C8H3BrF6 Formula Weight: 293.01 Flash Point: 110°(230°F) Refractive Index: 1.4270 Melting Point: -16° Boiling Point: 152-155° Beilstein Reference: 2123669 MDL No.: MFCD00000381
Formel: C₈H₃BrF₆
MW: 293.01 g/mol
Kokpunkt: 152…155 °C
Smältpunkt: –16 °C
Densitet: 1.698
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000381
CAS nummer: 328-70-1
EINECS: 206-334-5