α,α'-Dibromo-p-xylene ≥97%
Leverantör: Thermo Fisher Scientific
Fara
A13939.09 A13939.18
A13939.09EA 825 SEK
A13939.09 A13939.18
α,α'-Dibromo-p-xylene ≥97%
α,α'-Dibromo-p-xylene
Purity: 97% CAS No.: 623-24-5 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 144-148° Boiling Point: 245° UN No.: 3261 MDL No.: MFCD00000182
Formel: C₈H₈Br₂
Kokpunkt: 245 °C
Smältpunkt: 144…148 °C
Densitet: 2,012
Storage Temperature: Omgivande
MDL Number: MFCD00000182
CAS nummer: 623-24-5
EINECS: 210-781-1
UN: 3261
ADR: 8,II