4,5-Dibromo-o-xylene ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2-Dibromo-4,5-dimethylbenzene, 4,5-Dibromo-1,2-dimethylbenzene
B22727.03 B22727.06
B22727.03EA 1075 SEK
B22727.03 B22727.06
4,5-Dibromo-o-xylene ≥97%
4,5-Dibromo-o-xylene
Purity: 97% CAS No.: 24932-48-7 Molecular Formula: C8H8Br2 Formula Weight: 263.97 Melting Point: 85-87° Beilstein Reference: 2206183
Formel: C₈H₈Br₂
MW: 263,96 g/mol
Smältpunkt: 85…87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082743
CAS nummer: 24932-48-7