2,3-Dimethylphenylboronic acid ≥98%
Leverantör: Thermo Scientific
Varning
B23942.03 B23942.06 B23942.14
B23942.03EA 620 SEK
B23942.03 B23942.06 B23942.14
2,3-Dimethylphenylboronic acid ≥98%
2,3-Dimethylphenylboronic acid
Purity: 98% CAS No.: 183158-34-1 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: ca 175° Beilstein Reference: 8050763
Formel: C₈H₁₁BO₂
MW: 149,98 g/mol
Smältpunkt: ∼175 °C
Storage Temperature: Omgivande
MDL Number: MFCD01863524
CAS nummer: 183158-34-1