1,3-Diphenylpropane ≥98%
Leverantör: Thermo Scientific
L08849.09 L08849.04
L08849.09EA 7250 SEK
L08849.09 L08849.04
1,3-Diphenylpropane ≥98%
1,3-Diphenylpropane
Purity: 98% CAS No.: 1081-75-0 Molecular Formula: C15H16 Formula Weight: 196.29 Refractive Index: 1.5600 Boiling Point: 158-159°/10mm MDL No.: MFCD00043574
Formel: (C₆H₅)CH₂CH₂CH₂(C₆H₅)
MW: 196,29 g/mol
Kokpunkt: 158…159 °C (10 mmHg)
Densitet: 0,980
Storage Temperature: Omgivande
MDL Number: MFCD00043574
CAS nummer: 1081-75-0
EINECS: 214-101-4