Tymolblått natriumsalt ACS
Leverantör: Thermo Fisher Scientific
Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium
420940100 420940250 420941000
ACRO420940100EA 905 SEK
ACRO420940100 ACRO420940250 ACRO420941000
Tymolblått natriumsalt ACS
Tymolblått natriumsalt
Formel: C₂₇H₂₉NaO₅S
MW: 488.58 g/mol
Smältpunkt: 283 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00151093
CAS nummer: 62625-21-2
EINECS: 263-650-6
Order Now
Specifikation testresultat
Appearance Dark orange to green-black to brown-black Powder
Infrared spectrum Conforms
Visual transition interval From pH 8.0 (yellow) to pH 9.2 (blue)
Clarity of solution Passes test