4-Biphenylylmethanol ≥98%
Leverantör: Thermo Scientific
Synonyms: Biphenyl-4-methanol, [1,1'-Biphenyl]-4-ylmethanol, 4-Phenylbenzyl alcohol, 4-Biphenylmethanol
A11876.09 A11876.18 A11876.30
A11876.09EA 1375 SEK
A11876.09 A11876.18 A11876.30
4-Biphenylylmethanol ≥98%
4-Biphenylylmethanol
Purity: 98+% CAS No.: 3597-91-9 Molecular Formula: C13H12O Formula Weight: 184.24 Melting Point: 98-104° Boiling Point: 184°/11mm MDL No.: MFCD00004660
Formel: C₁₃H₁₂O
MW: 184,24 g/mol
Kokpunkt: 184 °C (11 mmHg)
Smältpunkt: 98…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004660
CAS nummer: 3597-91-9
EINECS: 222-745-2