Benzhydryl alcohol 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Diphenylcarbinol, Benzhydrol, Diphenylmethanol
105391000 105395000
ACRO105391000EA 605 SEK
ACRO105391000 ACRO105395000
Benzhydryl alcohol 99%
Benzhydryl alcohol
Formel: C₁₃H₁₂O
Kokpunkt: 297 °C (1013 hPa)
Smältpunkt: 65…68 °C
Densitet: 1,054 g/cm³ (80 °C)
Flampunkt: 160 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004488
CAS nummer: 91-01-0
EINECS: 202-033-8
Merck Index: 12,01121
Order Now
Specifikation testresultat
Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 65 to 68 °C
Water ≤0.5 % (K.F.)
Loss on drying ≤0.5 % (55 °C)
GC ≥98.5 %
Lead (Pb) ≤10 ppm
Nickel (Ni) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Insoluble matter ≤0.01 %