Benzil ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,2-Diphenyl-1,2-ethanedione
A12579.0E A12579.22 A12579.36
A12579.0EEA 6275 SEK
A12579.0E A12579.22 A12579.36
Benzil ≥98%
Benzil
Purity: 98% CAS No.: 134-81-6 Molecular Formula: C14H10O2 Formula Weight: 210.23 Melting Point: 94-96° Boiling Point: 346-348° Merck Reference: 14,1078 MDL No.: MFCD00003080
Formel: (C₆H₅)C(O)C(O)(C₆H₅)
Kokpunkt: 346…348 °C
Smältpunkt: 94…96 °C
Densitet: 1,521
Storage Temperature: Omgivande
MDL Number: MFCD00003080
CAS nummer: 134-81-6
EINECS: 205-157-0
Merck Index: 14,1078