2,4-Dihydroxybenzophenone ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (2,4-Dihydroxyphenyl)(phenyl)methanone, DHBP
A13749.0E A13749.22 A13749.36
A13749.0EEA 15175 SEK
A13749.0E A13749.22 A13749.36
2,4-Dihydroxybenzophenone ≥99%
2,4-Dihydroxybenzophenone
Purity: 99% CAS No.: 131-56-6 Molecular Formula: C13H10O3 Formula Weight: 214.22 Flash Point: 125°(257°F) Melting Point: 144-147° Boiling Point: 194°/1mm Beilstein Reference: 1311566 Merck Reference: 14,1106 MDL No.: MFCD00002277
Formel: C₁₃H₁₀O₃
MW: 214.22 g/mol
Kokpunkt: 194 °C (1 mmHg)
Smältpunkt: 144…147 °C
Densitet: 1.320
Flampunkt: 125 °C (257 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002277
CAS nummer: 131-56-6
EINECS: 205-029-4
Merck Index: 13,01107