4-Hydroxybenzophenone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: (4-Hydroxyphenyl)(phenyl)methanone, p-Hydroxybenzophenone
A15096.0B A15096.18 A15096.30
A15096.0BEA 12075 SEK
A15096.0B A15096.18 A15096.30
4-Hydroxybenzophenone ≥98%
4-Hydroxybenzophenone
Purity: 98% CAS No.: 1137-42-4 Molecular Formula: C13H10O2 Formula Weight: 198.22 Melting Point: 132-135° Boiling Point: 260-262°/24mm MDL No.: MFCD00002355
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
Kokpunkt: 260…262 °C (24 mmHg)
Smältpunkt: 132…135 °C
Densitet: 1,133 g/cm³ (172 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002355
CAS nummer: 1137-42-4
EINECS: 214-507-1