Bromofenolblått Reag. Ph. Eur. indicator, Fluka™
Leverantör: Honeywell Chemicals
Varning
Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
32712-5G 32712-25G
32712-5GEA 735 SEK
32712-5G 32712-25G
Bromofenolblått Reag. Ph. Eur. indicator, Fluka™
Bromofenolblått
Formel: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Smältpunkt: 204 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005875
CAS nummer: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429
Order Now
Specifikation testresultat
Indicator solution complying
Test of sensitivity complying
Change range (pH 2,8-4,4) complying
UV spectrum complying