Bromtymolblått sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Leverantör: Honeywell Chemicals
Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
32714-1KG 32714-5G 32714-25G
32714-1KGEA 28825 SEK
32714-1KG 32714-5G 32714-25G
Bromtymolblått sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Bromtymolblått
Formel: C₂₇H₂₈Br₂O₅S
MW: 624,39 g/mol
Smältpunkt: 204…202 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005872
CAS nummer: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430
Order Now
Specifikation testresultat
Suitability as indicator complying
Test of sensitivity complying
Clearness of the solution complying
Change Range (pH6,0-7,4) complying
Curve course (UV) complying