Sodium toluene-4-sulphonate
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: p-Toluenesulphonic acid sodium salt, Sodium p-toluenesulphonate, Toluene-4-sulphonic acid sodium salt
421231000 421235000
ACRO421231000EA 1175 SEK
ACRO421231000 ACRO421235000
Sodium toluene-4-sulphonate
Sodium toluene-4-sulphonate
Formel: C₇H₇NaO₃S
MW: 194.19 g/mol
Smältpunkt: >400 °C
Flampunkt: >500 °C
MDL Number: MFCD00798566
CAS nummer: 657-84-1
EINECS: 211-522-5
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration after Ion exchange ≥97.5 %
Titration with TBAH ≥98.0 %