p-Toluenesulphonyl hydrazide 98
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Methylbenzenesulphonylhydrazide, 4-Toluenesulphonyl hydrazide, 4-Methylbenzenesulphonhydrazide, 4-Methylbenzenesulphonic acid hydrazide, 4-Methylbenzenesulphonohydrazide, Toluene-4-sulphonohydrazide
A13529.14 A13529.22 A13529.36
A13529.14EA 890 SEK
A13529.14 A13529.22 A13529.36
p-Toluenesulphonyl hydrazide 98
p-Toluenesulphonyl hydrazide
Purity: 98% CAS No.: 1576-35-8 Molecular Formula: C7H10N2O2S Formula Weight: 186.23 Melting Point: 108-111° Beilstein Reference: 610130 UN No.: 3226 MDL No.: MFCD00007588
Formel: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Kokpunkt: 332
Smältpunkt: 108…111 °C
Densitet: 1.4
Storage Temperature: Omgivande
MDL Number: MFCD00007588
CAS nummer: 1576-35-8
EINECS: 216-407-3
UN: 3226
ADR: 4.1 II