Toluen-4-sulfonamid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: PTSA, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide
147690010 147692500 147690050
ACRO147690010EA 1400 SEK
ACRO147690010 ACRO147692500 ACRO147690050
Toluen-4-sulfonamid 99%
Toluen-4-sulfonamid
Formel: C₇H₉NO₂S
MW: 171,22 g/mol
Kokpunkt: 221 °C (10 mmHg)
Smältpunkt: 136…140 °C
Flampunkt: 202 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011692
CAS nummer: 70-55-3
EINECS: 200-741-1
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥98.5 %