p-Toluensulfonylklorid 98
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-p-sulphonyl chloride
A14547.0B A14547.0I A14547.30 A14547.36
A14547.0BEA 2075 SEK
A14547.0B A14547.0I A14547.30 A14547.36
p-Toluensulfonylklorid 98
p-Toluensulfonylklorid
Purity: 98% CAS No.: 98-59-9 Molecular Formula: C7H7ClO2S Formula Weight: 190.65 Flash Point: 128°(262°F) Melting Point: 67-70° Boiling Point: 146°/15mm Solubility: Soluble in alcohol, benzene, and ether. Insoluble in water Beilstein Reference: 607898 Merck Reference: 14,9534 UN No.: 3261 MDL No.: MFCD00007450
Formel: C₇H₇ClO₂S
MW: 190.65 g/mol
Kokpunkt: 146 °C (15 mmHg)
Smältpunkt: 67…70 °C
Densitet: 1.35
Flampunkt: 128 °C (262 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007450
CAS nummer: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8 II
Merck Index: 13,09612