4-Hexylresorcinol ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Hexylresorcinol, 4-Hexylbenzene-1,3-diol
A12552.06 A12552.14 A12552.22
A12552.06EA 515 SEK
A12552.06 A12552.14 A12552.22
4-Hexylresorcinol ≥99%
4-Hexylresorcinol
Purity: 99% CAS No.: 136-77-6 Molecular Formula: C12H18O2 Formula Weight: 194.27 Flash Point: 100°(230°F) Melting Point: 64-67° Boiling Point: 333-335° Beilstein Reference: 2048312 Merck Reference: 14,4712 MDL No.: MFCD00002284
Formel: C₁₂H₁₈O₂
MW: 194,27 g/mol
Kokpunkt: 333…335 °C
Smältpunkt: 64…67 °C
Flampunkt: >100 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002284
CAS nummer: 136-77-6
EINECS: 205-257-4
Merck Index: 14,4712