4-tert-Butylkatekol 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol
154351000 154355000 154350025 154350100
ACRO154351000EA 565 SEK
ACRO154351000 ACRO154355000 ACRO154350025 ACRO154350100
4-tert-Butylkatekol 99%
4-tert-Butylkatekol
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
Kokpunkt: 285 °C (760 mmHg)
Smältpunkt: 53…56 °C
Densitet: 1,049 g/cm³
Flampunkt: 129 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002201
CAS nummer: 98-29-3
EINECS: 202-653-9
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White to cream Flakes
Infrared spectrum Conforms
Melting point 53 to 56 °C
Water ≤0.3 % (K.F.)
GC ≤0.7 % Catechol