3-tert-Butylphenol 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
Synonyms:
m-tert-Butylphenol
B0731-25G
B0731-500G
TCIAB0731-25GEA
470
SEK
TCIAB0731-25G
TCIAB0731-500G
3-tert-Butylphenol 98,0 GC_ANALYSMETOD
3-tert-Butylphenol
Formel:
C₁₀H₁₄O MW: 150.22 g/mol Kokpunkt: 132°C Smältpunkt: 43 |
MDL Number:
MFCD00002300 CAS nummer: 585-34-2 EINECS: 209-553-4 UN: 2430 ADR: 8 III |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...