3,4-Dimethoxybenzyl alcohol 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Veratryl alcohol, (3,4-Dimethoxyphenyl)methanol
115490250 115491000 115495000
ACRO115490250EA 710 SEK
ACRO115490250 ACRO115491000 ACRO115495000
3,4-Dimethoxybenzyl alcohol 96%
3,4-Dimethoxybenzyl alcohol
Formel: C₉H₁₂O₃
Kokpunkt: 296…297 °C (732 mmHg)
Smältpunkt: 22 °C
Densitet: 1,15 g/cm³
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004638
CAS nummer: 93-03-8
EINECS: 202-212-0
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to golden Viscous liquid
Infrared spectrum Conforms
GC ≥97.5 %