4-Metyl-m-fenylendiamin 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Metyl-1,3-benzendiamin
146600010 146600250 146602500
ACRO146600010EA 10025 SEK
ACRO146600010 ACRO146600250 ACRO146602500
4-Metyl-m-fenylendiamin 98%
4-Metyl-m-fenylendiamin
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Kokpunkt: 283…285 °C (760 mmHg)
Smältpunkt: 97…101 °C
Densitet: 1,045 g/cm³ (20 °C)
Flampunkt: 149 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007804
CAS nummer: 95-80-7
EINECS: 202-453-1
UN: 1709
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Dark grey to brown Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 95 to 102 °C
HPLC ≥97.5 %