3-Methoxybenzylamine 98%
Leverantör: Thermo Scientific
Fara
Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole
170550100 170550250
ACRO170550100EA 1350 SEK
ACRO170550100 ACRO170550250
3-Methoxybenzylamine 98%
3-Methoxybenzylamine
Formel: C₈H₁₁NO
MW: 137,18 g/mol
Kokpunkt: 140 °C (37 mmHg)
Densitet: 1,07 g/cm³
Flampunkt: >110 °C
MDL Number: MFCD00008115
CAS nummer: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %