2-Isopropylphenyl isothiocyanate 96%
Leverantör: Thermo Scientific
Fara
A19448.06 A19448.14
A19448.06EA 2275 SEK
A19448.06 A19448.14
2-Isopropylphenyl isothiocyanate 96%
2-Isopropylphenyl isothiocyanate
Purity: 96% CAS No.: 36176-31-5 Molecular Formula: C10H11NS Formula Weight: 177.27 Refractive Index: 1.6050 Boiling Point: 131-132°/10mm UN No.: 2922 MDL No.: MFCD00022053
Formel: C₁₀H₁₁NS
MW: 177,27 g/mol
Kokpunkt: 131…132 °C (10 mmHg)
Densitet: 1,050
Storage Temperature: Omgivande
MDL Number: MFCD00022053
CAS nummer: 36176-31-5
ADR: 8,III