Benzyl isothiocyanate ≥98%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Benzylisothiocyanate

A15008.06 A15008.14 A15008.22
A15008.06EA 760 SEK
A15008.06 A15008.14 A15008.22
Benzyl isothiocyanate ≥98%
Benzyl isothiocyanate

Purity: 98% CAS No.: 622-78-6 Molecular Formula: C6H5CH2NCS Formula Weight: 149.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6030 Odour: Pungent Boiling Point: 242-243° Beilstein Reference: 386135 UN No.: 2922 MDL No.: MFCD00004819

Formel: C₈H₇NS
MW: 149.22 g/mol
Kokpunkt: 242…243 °C
Smältpunkt: 24
Densitet: 1.125
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00004819
CAS nummer: 622-78-6
EINECS: 210-753-9
UN: 2922
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR