2-Phenylethyl isothiocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: β-Phenethyl isothiocyanate, 2-Phenethyl isothiocyanate, 2-Isothiocyanatoethylbenzene, Phenethyl isothiocyanate
A15123.06 A15123.14
A15123.06EA 1225 SEK
A15123.06 A15123.14
2-Phenylethyl isothiocyanate ≥98%
2-Phenylethyl isothiocyanate
Purity: 98% CAS No.: 2257-09-2 Molecular Formula: C6H5(CH2)2NCS Formula Weight: 163.24 Flash Point: 110°(230°F) Refractive Index: 1.5888 Odour: Pungent Boiling Point: 139-140°/11mm Beilstein Reference: 2084162 UN No.: 2922 MDL No.: MFCD00004821
Formel: C₆H₅CH₂CH₂NCS
MW: 163.24 g/mol
Kokpunkt: 139…140 °C (11 mmHg)
Densitet: 1.090
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004821
CAS nummer: 2257-09-2
EINECS: 218-855-5
ADR: 8 III