2-Ethylphenyl isocyanate

AC31420DA
MAYBAC31420DAEA 0 SEK
MAYBAC31420DA
2-Ethylphenyl isocyanate
2-Ethylphenyl isocyanate
Formel: C₉H₉NO
MW: 147,18 g/mol
Kokpunkt: 56 °C (3 mmHg)
Densitet: 1,04 g/cm³ (20 °C)
MDL Number: MFCD00002013
CAS nummer: 40411-25-4
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR