Benzyl(dimetyl)amin 99%
Leverantör: Thermo Scientific
Fara
Synonyms: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA
159751000 159755000 159750025 159750051
ACRO159751000EA 425 SEK
ACRO159751000 ACRO159755000 ACRO159750025 ACRO159750051
Benzyl(dimetyl)amin 99%
Benzyl(dimetyl)amin
Formel: C₆H₅CH₂N(CH₃)₂
MW: 135,21 g/mol
Kokpunkt: 183…184 °C (760 mmHg)
Smältpunkt: –75 °C
Densitet: 0,9 g/cm³
Flampunkt: 54 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008329
CAS nummer: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=6f09c2612c86a9aa5e15ab2e99c6ad8cd75aa291&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5020 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %
Filt.over 0.2 micron PFTE Filtered