α,α,α-Trifluoro-p-toluidine 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A
208910100 208910500
ACRO208910100EA 1125 SEK
ACRO208910100 ACRO208910500
α,α,α-Trifluoro-p-toluidine 99%
α,α,α-Trifluoro-p-toluidine
Formel: C₇H₆F₃N
MW: 161.13 g/mol
Kokpunkt: 83 °C (12 mmHg)
Smältpunkt: 5…8 °C
Densitet: 1.296 g/cm³
Flampunkt: 86
Storage Temperature: Omgivande
MDL Number: MFCD00064396
CAS nummer: 455-14-1
EINECS: 207-236-5
UN: 2810
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4860 (20°C, 589 nm)
GC ≥98.5 %