2-Fluoro-5-(trifluoromethyl)aniline ≥97%
Leverantör: Thermo Fisher Scientific
|
Fara
|
Purity: 97%
CAS No.: 535-52-4
Molecular Formula: C7H5F4N
Formula Weight: 179.12
Flash Point: 70°(158°F)
Refractive Index: 1.4610
Odour: Characteristic
Boiling Point: 210°
Beilstein Reference: 1950800
UN No.: 2810
MDL No.: MFCD00007653
Formel:
C₇H₅F₄N Kokpunkt: 210 °C Densitet: 1,378 Flampunkt: 70 °C (158 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00007653 CAS nummer: 535-52-4 UN: 2810 ADR: 6.1,III |