4-(Trifluoromethyl)phenyl isocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
L12576.03 L12576.06
L12576.03EA 830 SEK
L12576.03 L12576.06
4-(Trifluoromethyl)phenyl isocyanate ≥98%
4-(Trifluoromethyl)phenyl isocyanate
Purity: 98% CAS No.: 1548-13-6 Molecular Formula: C8H4F3NO Formula Weight: 187.12 Storage Temperature: 2°C to 8°C Flash Point: 70°(158°F) Refractive Index: 1.4710 Odour: Pungent Boiling Point: 91-94°/35mm Beilstein Reference: 881041 UN No.: 2285 MDL No.: MFCD00002028
Formel: C₈H₄F₃NO
Kokpunkt: 91…94 °C (35 mmHg)
Smältpunkt: 58…59 °C
Densitet: 1,310
Flampunkt: 70 °C (158 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00002028
CAS nummer: 1548-13-6
EINECS: 216-284-6
UN: 2285
ADR: 6.1,II