α,α,α-Trifluoro-m-toluoyl chloride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride
312940050 312940250
ACRO312940050EA 760 SEK
ACRO312940050 ACRO312940250
α,α,α-Trifluoro-m-toluoyl chloride 98%
α,α,α-Trifluoro-m-toluoyl chloride
Formel: C₈H₄ClF₃O
MW: 208,57 g/mol
Kokpunkt: 184…186 °C (750 mmHg)
Densitet: 1,38 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000680
CAS nummer: 2251-65-2
EINECS: 218-844-5
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4780 (20°C, 589 nm)
GC ≥97.5 %