2',4'-Difluoroacetophenone 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-(2,4-Difluorophenyl)ethan-1-one
246770100 246770500
ACRO246770100EA 1600 SEK
ACRO246770100 ACRO246770500
2',4'-Difluoroacetophenone 99%
2',4'-Difluoroacetophenone
Formel: C₈H₆F₂O
MW: 156,13 g/mol
Kokpunkt: 80…81 °C (25 mmHg)
Densitet: 1,23 g/cm³
Flampunkt: 66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151261
CAS nummer: 364-83-0
EINECS: 206-667-6
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4890 (20°C, 589 nm)
GC ≥98.5 %