α,α,α-Trifluoro-2-tolualdehyde 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-(Trifluoromethyl)benzaldehyde
232680250 232681000
ACRO232680250EA 1675 SEK
ACRO232680250 ACRO232681000
α,α,α-Trifluoro-2-tolualdehyde 97%
α,α,α-Trifluoro-2-tolualdehyde
Formel: C₈H₅F₃O
MW: 174.12 g/mol
Kokpunkt: 70…71 °C (16 mmHg)
Smältpunkt: –40 °C
Densitet: 1.32 g/cm³
Flampunkt: 61 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003337
CAS nummer: 447-61-0
EINECS: 207-185-9
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %