4-(Trifluoromethyl)phenylboronic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid
359765000 359760050
ACRO359765000EA 520 SEK
ACRO359765000 ACRO359760050
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid
Formel: C₇H₆BF₃O₂
MW: 189.93 g/mol
Smältpunkt: 245…250 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00151855
CAS nummer: 128796-39-4
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %