3-(Trifluoromethyl)phenylboronic acid 97+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane
359750010 359750050
ACRO359750010EA 535 SEK
ACRO359750010 ACRO359750050
3-(Trifluoromethyl)phenylboronic acid 97+%
3-(Trifluoromethyl)phenylboronic acid
Formel: C₇H₆BF₃O₂
MW: 189.93 g/mol
Smältpunkt: 160…165 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00151854
CAS nummer: 1423-26-3
Order Now
Specifikation testresultat
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 161 to 167 °C
HPLC ≥97.0 %