2-(Trifluoromethyl)phenylboronic acid ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid
B24343.03 B24343.06 B24343.14
B24343.03EA 925 SEK
B24343.03 B24343.06 B24343.14
2-(Trifluoromethyl)phenylboronic acid ≥97%
2-(Trifluoromethyl)phenylboronic acid
Purity: 97% CAS No.: 1423-27-4 Molecular Formula: C7H6BF3O2 Formula Weight: 189.93 Melting Point: 108-110° Beilstein Reference: 2974465
Formel: C₇H₆BF₃O₂
MW: 189.93 g/mol
Smältpunkt: 108…110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00236059
CAS nummer: 1423-27-4